Learn More
Kieselguhr, Pure, Washed with Acid, Fisher Chemical™
Brand: Fisher Chemical K/0100/60
241.99 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description

Specifications
Solid | |
1 kg | |
61790-53-2 | |
O2Si |
Gray | |
9 to 10 | |
Plastic Keg | |
132803 |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Kieselguhr
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H373-May cause damage to organs.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.