missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Silicone Stoppers, Single Hole
Kimble™ Silicone Stoppers are general purpose single-hole silicone rubber stoppers. They are steam autoclavable and an opaque yellowish-white in color. For use with Kimble™ Kontes™ Filtration Assemblies.
£20.00 - £25.10
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
15269833
|
DWK Life Sciences
953715-0501 |
No. 5 |
£20.00
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
15249833
|
DWK Life Sciences
953763-0801 |
No. 8 |
£25.10
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
|||||
Description
- Product Type: Stopper
- Material: Silicone Rubber
- Color: White/Yellow
- Height: 25 mm
- Autoclavable: Yes
- Use with: Kimble™ Kontes™ Filtration Assemblies
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title