missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimtech™ Process Wipers

Designed for specific tasks in controlled areas. Kimtech™ Process Wipers are ideal for repeated, lint-free cleaning.

Brand:  Kimberly-Clark™ 7643

154.38 GBP valid until 2024-12-31
Use promo code "24046" to get your promotional price.


 View more versions of this product

Product Code. 10748444

  • £334.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Ideal for repeated, lint-free cleaning with chemicals and solvents
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Process Wiper
38 cm
34 cm
White
Repeat-use cleaning with chemicals and solvents, without leaving lint
Non-sterile
15 in.
13.6 in.
Polypropylene
38 x 34 cm (15 x 13.4 in.)
1-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers