missing translation for 'onlineSavingsMsg'
Learn More
Kjeldahl 4% Boric Acid Solution, without Indicator, Fisher Chemical™ Product Code.: 15968866

Kjeldahl 4% Boric Acid Solution, without Indicator, Fisher Chemical™

Product Code. 15968866
1 L
Click to view available options
Quantity:
1 L
5 L
This item is not returnable. View return policy

Product Code. 15968866

Brand: Fisher Chemical K/0205/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Premade 4% boric acid solution for nitrogen determination by Kjeldahl method.

TRUSTED_SUSTAINABILITY

Specifications

Packaging HDPE Plastic Bottle
Assay Percent Range 3.9 - 4.1%
Quantity 1 L
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kjeldahl 4% Boric Acid Solution, without Indicator, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.