missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klercide™ 70/30 IPA Blended with WFI
70% isopropyl alcohol blended with Water for Injection (WFI) with endotoxin level guaranteed to be less than 0.25 EU/mL
Brand: Ecolab™ 3078550
This item is not returnable.
View return policy
Description
- The alcohol blend is 0.2 micron filtered then filled and triple bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
Specifications
Sterile IPA Blended with WFI Stream Aerosol | |
24/Pk. | |
360 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Klercide 70/30 IPA Aerosol
Signal Word
- Danger
Hazard Category
- Flammable and non-flammable aerosols category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H222-Extremely flammable aerosol.
- H229-Pressurised container: May burst if heated.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P211-Do not spray on an open flame or other ignition source.
- P251-Do not pierce or burn, even after use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl Alcohol