missing translation for 'onlineSavingsMsg'
Learn More

Ecolab™ Klercide™ 70/30 IPA Blended with WFI

70% isopropyl alcohol blended with Water for Injection (WFI) with endotoxin level guaranteed to be less than 0.25 EU/mL

£685.00 - £1120.00

Specifications

Quantity 24/Pk.
Volume (Metric) 360 mL
View More Specs

Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15346088
View Documents
Ecolab™
3078580
Blended with WFI Aerosol This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15392438
View Documents
Ecolab™
3078550
Sterile IPA Blended with WFI Stream Aerosol This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • The alcohol blend is 0.2 micron filtered then filled and triple bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
Specifications

Specifications

24/Pk.
360 mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Klercide 70/30 IPA Aerosol

Signal Word
  • Danger

Hazard Category
  • Flammable and non-flammable aerosols category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H222-Extremely flammable aerosol.
  • H229-Pressurised container: May burst if heated.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P211-Do not spray on an open flame or other ignition source.
  • P251-Do not pierce or burn, even after use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.

Supplemental information
  • MIXTURE LIST-Contains : Isopropyl Alcohol

SDS
Documents

Documents

Product Certifications