missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klercide™ 70/30 Isopropyl Alcohol Blended with WFI
For use in effective disinfection procedure. Ecolab™ Klercide™ 70/30 Isopropyl Alcohol Blended with WFI is sterilized by gamma irradiation.
Brand: Ecolab™ 3078540
This item is not returnable.
View return policy
Description
- Available as either 70/30 Isopropyl Alcohol (IPA) or Denatured Ethanol blended with water for injection (WFI) or deionized water
- Also available as 60% Isopropyl alcohol or Propan-1-ol and Denatured Ethanol, both blended with WFI
- Use pharma grade alcohol in blend
- Cleanroom processed, 0.2 micron filtered and terminally sterilized by gamma irradiation
- Available as spray systems fitted with SteriShield Delivery System, which ensures sterility of contents is preserved throughout use, aerosols, stream aerosol or in 5L capped container
- Available with complete supporting documentation
Specifications
Alcohol with WFI | |
0.2 μm | |
6/Cs. |
Sterile | |
Suitable for cleanroom use | |
1 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Klercide 70/30 IPA
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl Alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction