missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolab™ Klercide™ 70/30 Isopropyl Alcohol Blended with WFI
For use in effective disinfection procedure. Ecolab™ Klercide™ 70/30 Isopropyl Alcohol Blended with WFI is sterilized by gamma irradiation.
£745.00 - £1398.94
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15306088
|
Ecolab™
3078510 |
5 L | N/A | ||||||
16479139
|
Ecolab™
3078510 |
5 L | N/A | ||||||
15326088
|
Ecolab™
3078540 |
1 L | N/A | ||||||
15336088
|
Ecolab™
3078560 |
500 mL |
£1398.94
Pack of 12 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Available as either 70/30 Isopropyl Alcohol (IPA) or Denatured Ethanol blended with water for injection (WFI) or deionized water
- Also available as 60% Isopropyl alcohol or Propan-1-ol and Denatured Ethanol, both blended with WFI
- Use pharma grade alcohol in blend
- Cleanroom processed, 0.2 micron filtered and terminally sterilized by gamma irradiation
- Available as spray systems fitted with SteriShield Delivery System, which ensures sterility of contents is preserved throughout use, aerosols, stream aerosol or in 5L capped container
- Available with complete supporting documentation
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Klercide 70/30 IPA
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl Alcohol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title