Learn More
Ecolab™ Klercide™ 70/30 Isopropyl Alcohol Blended with WFI
For use in effective disinfection procedure. Ecolab™ Klercide™ 70/30 Isopropyl Alcohol Blended with WFI is sterilized by gamma irradiation.
Brand:  Ecolab™ 3078510
 View more versions of this product
Product Code. 16479139
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Available as either 70/30 Isopropyl Alcohol (IPA) or Denatured Ethanol blended with water for injection (WFI) or deionized water
  • Also available as 60% Isopropyl alcohol or Propan-1-ol and Denatured Ethanol, both blended with WFI
  • Use pharma grade alcohol in blend
  • Cleanroom processed, 0.2 micron filtered and terminally sterilized by gamma irradiation
  • Available as spray systems fitted with SteriShield Delivery System, which ensures sterility of contents is preserved throughout use, aerosols, stream aerosol or in 5L capped container
  • Available with complete supporting documentation
Specifications
Specifications
Filtered Alcohol
Filtered
Disinfection procedure
5 L