Learn More
Ecolab™ Klercide™ Sporicidal Low Residue Peroxide Blended With WFI
A very low residue blend of high grade 6% hydrogen peroxide and Water for Injection (WFI) with endotoxin level guaranteed to be less than 0.25 EU/mL
Brand: Ecolab™ 3079120
This item is not returnable.
View return policy
Description
- The biocide blend is 0.2 micron filtered and sterile filled under Grade A laminar flow and triple bagged in a Grade B cleanroom.
- The product is a sterile ready-to-use rotational sporicide with very low residues for small and large surfaces.
Specifications
Blended with WFI | |
Cap | |
4 Pk. | |
5 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Klercide Sporicidal Low Residue Peroxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Hydrogen peroxide