Learn More
Ecolab™ Klercide™ Sporicidal Low Residue Peroxide Blended With WFI
A very low residue blend of high grade 6% hydrogen peroxide and Water for Injection (WFI) with endotoxin level guaranteed to be less than 0.25 EU/mL
£541.00 - £1170.00
Specifications
Description Blended with WFI
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15356108
View Documents
Ecolab™
3079120
5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15679185
View Documents
Ecolab™
3079100
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • The biocide blend is 0.2 micron filtered and sterile filled under Grade A laminar flow and triple bagged in a Grade B cleanroom.
  • The product is a sterile ready-to-use rotational sporicide with very low residues for small and large surfaces.
Specifications
Specifications
Blended with WFI
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Klercide Sporicidal Low Residue Peroxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen peroxide