Learn More
Ecolab Klerwipe 70/30 IPA Pouch Wipe Blended with Water for Injection
These sterile Klerwipe 70/30 IPA knitted polyester wipes are impregnated with 0.2 micron filtered IPA blended with Water for Injection
Brand:  Ecolab 6601007
Additional Details : CAS Number : 67-63-0 Weight : 3.10700kg
 View more versions of this product
Product Code. 17815507
  • £630.00 / Pack of 150
Estimated Shipment: 01-08-2024
to see stock.
Description
Description
  • BPR approved
  • High grade solution for rapid controlled disinfection of small surfaces, equipment and production components
  • Impregnated with 70/30 v/v IPA blended with Water for Injection. The alcohol blend is 0.2 micron filtered, then filled and triple bagged in a Grade C (ISO Class 7) cleanroom and irradiated using a validated process at no less than 25 kGy
  • Low residue product, contains no chemical binders and as such, after the desired contact time, when the alcohol has evaporated, there are no residues left on the surface
  • Made of high quality, pre-laundered knitted polyester with ultrasonically sealed edges
  • Has an Endotoxin level of less than 1 EU per wipe
  • Provides a reduced risk of inhalation
  • Each pouch contains just 10 wipes – ideal for sessional use without compromising sterility
  • Provides high absorbency and consistent application
  • The packs feature Klertear for easy opening, even while wearing gloves
  • Certificates of Analysis are available for every batch
Specifications
Specifications
Wipe
300 mm
Sterile
300 mm
Polyester