Learn More
Ecolab Klerwipe 70/30 IPA Pouch Wipe Blended with Water for Injection
These sterile Klerwipe 70/30 IPA knitted polyester wipes are impregnated with 0.2 micron filtered IPA blended with Water for Injection
Brand:  Ecolab 6601003
Additional Details : CAS Number : 67-63-0 Weight : 1.84300kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
 View more versions of this product
Product Code. 17835507
  • £570.00 / Pack of 200
Estimated Shipment: 01-08-2024
to see stock.
Description
Description
  • BPR approved
  • High grade solution for rapid controlled disinfection of small surfaces, equipment and production components
  • Impregnated with 70/30 v/v IPA blended with Water for Injection. The alcohol blend is 0.2 micron filtered, then filled and triple bagged in a Grade C (ISO Class 7) cleanroom and irradiated using a validated process at no less than 25 kGy
  • Low residue product, contains no chemical binders and as such, after the desired contact time, when the alcohol has evaporated, there are no residues left on the surface
  • Made of high quality, pre-laundered knitted polyester with ultrasonically sealed edges
  • Has an Endotoxin level of less than 1 EU per wipe
  • Provides a reduced risk of inhalation
  • Each pouch contains just 10 wipes – ideal for sessional use without compromising sterility
  • Provides high absorbency and consistent application
  • The packs feature Klertear for easy opening, even while wearing gloves
  • Certificates of Analysis are available for every batch
Specifications
Specifications
Wipe
200 mm
Sterile
200 mm
Polyester
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • X200 70/30 IPA 20X10
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Isopropyl Alcohol