missing translation for 'onlineSavingsMsg'
Learn More

Ecolab Klerwipe Sterile Low Particulate Dry Wipe

Klerwipe Sterile Low Particulate Dry Wipes are binder free hydroentangled polyester/cellulose wipes. Each pack, featuring Klertear, is packed in a Grade C cleanroom before being terminally sterilized by gamma irradiation.

Brand:  Ecolab 3035530

Additional Details : Weight : 1.70000kg

 View more versions of this product

Product Code. 17516526

  • £221.00 / Pack of 100
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Double bagged for ease of transfer
  • Compatible with the Klercide range of Alcohols and Biocides
  • Suitable for use in controlled environments such as isolators and laminar flow cabinets
  • All packaging is designed to minimize the generation of particles and features Klertear, which makes the packs easy to open even when wearing gloves
  • Cost effective – packed in small quantities to avoid the need to compromise the sterility of a pack.
Specifications

Specifications

Dry Wipes
400 mm
700 x 400 mm
Sterile
700 mm
Polyester Cellulose
Cleaning and Disinfection Procedures
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers