Learn More
KLF2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23384-1-AP-20UL
This item is not returnable.
View return policy
Description
The KLF2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KLF2. This antibody recognizes human, mouse antigen. The KLF2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
KLF2 | |
1000 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse | |
150 kDa | |
IgG |
Western Blot, ELISA | |
KLF2, Krueppel like factor 2, Kruppel like factor 2 (lung), LKLF, Lung krueppel like factor | |
20 μL | |
10365 | |
Store at -20°C. Stable for one year after shipment. | |