missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£155.00 - £404.20
Specifications
Antigen | KLF2 |
---|---|
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Form | Liquid |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18078185
|
Proteintech
23384-1-AP-150UL |
150 μL |
£404.20
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18037895
|
Proteintech
23384-1-AP-20UL |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The KLF2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KLF2. This antibody recognizes human, mouse antigen. The KLF2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
KLF2 | |
Western Blot, ELISA | |
Rabbit | |
10365 | |
IgG | |
Store at -20°C. Stable for one year after shipment. |
1000 μg/mL | |
Liquid | |
Human, Mouse | |
KLF2, Krueppel like factor 2, Kruppel like factor 2 (lung), LKLF, Lung krueppel like factor | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title