missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Amber 203 Serum Vials
Autoclavable; ideally suited for the handling, and storage of a variety of liquids including reagents, vaccines, blood plasma, culture media, chromatography.
Brand: DWK Life Sciences 62421D-30
Additional Details : Weight : 0.63000kg
Description
- Serum vial with an aluminum seal finish
- Comes without a closure and is lighter in weight than molded bottles
- Manufactured from amber 203 borosilicate glass
- Enhanced by the most popular GPI aluminum seal finishes, this line offers a variety of choices
Specifications
Amber | |
Borosilicate Glass | |
Ideally suited for the handling, and storage of a variety of liquids including reagents, vaccines, blood plasma, culture media, chromatography samples | |
8 dr. | |
86 mm |
30 mm | |
30 mL | |
Aluminum | |
Autoclavable | |
ASTM E438, Type I |