missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Amber 203 Serum Vials
Autoclavable; ideally suited for the handling, and storage of a variety of liquids including reagents, vaccines, blood plasma, culture media, chromatography.
£890.00
Specifications
Color | Amber |
---|---|
Material | Borosilicate Glass |
Material (Seal) | Aluminum |
Autoclavable | Autoclavable |
Certifications/Compliance | ASTM E438, Type I |
Product Code | Brand | Diameter (Metric) Outer | Volume (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Volume (Metric) | Height (Metric) | Price | Quantity & Availability | |||
17771078
|
DWK Life Sciences
62421D-30 |
30 mm | 30 mL | 86 mm |
£890.00
Pack of 576 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||
Description
- Serum vial with an aluminum seal finish
- Comes without a closure and is lighter in weight than molded bottles
- Manufactured from amber 203 borosilicate glass
- Enhanced by the most popular GPI aluminum seal finishes, this line offers a variety of choices
Specifications
Amber | |
Aluminum | |
ASTM E438, Type I |
Borosilicate Glass | |
Autoclavable |