Learn More
DWK Life Sciences Kimble™ Kontes™ Flask Inlet Adapter, Monier-Williams, 24/40 Joint
Component Parts of Monier-Williams Sulfites Distillation Apparatus which is used for the determination of free sulfite and a reproducible portion of the bound sulfites in food, such as the carbonyl addition products.
Brand:  DWK Life Sciences 513801-0000
Product Code. 15120526
  • £86.20 / Each
Explore more special offers
Description
Description
  • Flask Inlet Adapter
  • Standard Taper joint 24/40
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flask Inlet Adapter
24/40
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Flask Inlet Adapter, Monier-Williams, 24/40 Joint > Flask Inlet Adapter