missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask
Manufactured from Pyrex™ glass for high resistance to thermal shock
Brand: Pyrex 1080/02M
This item is not returnable.
View return policy
Description
- Complies with ISO 24450
- With wide neck to facilitate easy addition of solid samples etc
- Flat base for standing on benches and hotplates etc
- Manufactured from Pyrex™ glass for high resistance to thermal shock
- Excellent chemical durability for contamination free performance
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
ISO 24450 | |
Borosilicate Glass | |
Wide |
50 mL | |
75 mm | |
52 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask > Capacity: 50mL
Spot an opportunity for improvement?Share a Content Correction