missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Serum Vials KG-33 Borosilicate Glass with Aluminum Seal

Autoclavable Kimble™ serum vials are ideally suited for the handling, containment, and storage of a variety of liquids.

Brand:  DWK Life Sciences 62113D-2

600.16 GBP valid until 2024-12-31
Use promo code "21960" to get your promotional price.



Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10013444

  • £690.00 / Pack of 1440
Estimated Shipment: 21-08-2024
to see stock.
Add to basket

Description

Description

Autoclavable Kimble™ serum vials are ideally suited for the handling, containment, and storage of a variety of liquids.

  • Special design provides extra strength for freeze-drying applications
  • Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
  • Autoclavable
    Aluminum seals, stoppers and septa are available separately
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirement
Specifications

Specifications

A
15 mm
2 mL
Aluminum
Autoclavable
ASTM E438, Type I, Class A
Clear
Borosilicate Glass
Ideally suited for the handling, containment, and storage of a variety of liquids
0.5 dr.
40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers