missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Krebs-Ringer Solution, bicarbonate-buffered
Brand: Thermo Scientific Chemicals J67591.AP
83.88 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
500 mL | |
Plastic Bottle | |
Ambient Temperatures | |
Liquid |
RUO – Research Use Only