missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Krebs-Ringer Solution, bicarbonate-buffered

£96.48 - £136.68

Specifications

Packaging Plastic Bottle
Content And Storage Ambient Temperatures
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15418769
View Documents Promotion Details
Thermo Scientific Chemicals
J67591.AP
500 mL
£96.48
500mL
Please to purchase this item. Need a web account? Register with us today!
15428769
View Documents
Thermo Scientific Chemicals
J67591.K2
1 L
£136.68
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Plastic Bottle
Ambient Temperatures
Liquid
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only