missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Alanine, Cell Culture Reagent Product Code.: 15483069

Thermo Scientific Chemicals L-Alanine, Cell Culture Reagent

Product Code. 15483069
500 g
Click to view available options
Quantity:
50 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15483069

Brand: Thermo Scientific Chemicals J60279.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

Solubility
Soluble in water 166.5 g/L..

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acidShow MoreShow Less
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Melting Point ∽295°C (decomposition)
Color White
Beilstein 1720248
Merck Index 14,204
Solubility Information Soluble in water 166.5g/L..
Quantity 500 g
Formula Weight 89.09
Optical Rotation +14° (c=5 in 5N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Alanine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM20L010 Certificate TypeCertificate of Analysis Date22/01/2025 Catalog NumberJ60279.18,J60279.36,J60279.22
Lot NumberU27K046 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog NumberJ60279.18,J60279.36,J60279.22
Lot NumberR14K024 Certificate TypeCertificate of Analysis Date21/05/2024 Catalog NumberJ60279.18,J60279.36,J60279.22
Lot NumberM09K062 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog NumberJ60279.18,J60279.36,J60279.22
Lot NumberY07J005 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ60279.18,J60279.36,J60279.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Alanine, Cell Culture Reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.