Learn More
Thermo Scientific Chemicals L-Alanine, 99%
Description
L-Alanine plays a vital role in glucose-alanine cycle between tissue and liver. It is used for protein construction as well as involved in the metabolism of tryptophan and vitamin pyridoxine. It provides energy for muscles and central nervous system in order to strengthen the immune system. It helps in the metabolism of sugars and organic acids, and enhances the germination rates of Bacillus subtilis spores. It also displays a cholesterol-reducing effect in animals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Alanine plays a vital role in glucose-alanine cycle between tissue and liver. It is used for protein construction as well as involved in the metabolism of tryptophan and vitamin pyridoxine. It provides energy for muscles and central nervous system in order to strengthen the immune system. It helps in the metabolism of sugars and organic acids, and enhances the germination rates of Bacillus subtilis spores. It also displays a cholesterol-reducing effect in animals.
Solubility
Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | ∼295°C (decomposition) |
Assay Percent Range | 99% |
Beilstein | 1720248 |
Merck Index | 14,204 |
Solubility Information | Soluble in water and cold ethanol. Slightly soluble in pyridine. Insoluble in ether and acetone. |
Density | 1.432 |
Quantity | 500 g |
Formula Weight | 89.09 |
Optical Rotation | +14° (c=5 in 5N HCl) |
Percent Purity | 99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U26K013 | Certificate of Analysis | 06/09/2024 | A15804.22,A15804.14,A15804.36 |
U02K043 | Certificate of Analysis | 16/08/2024 | A15804.22,A15804.14,A15804.36 |
U01K040 | Certificate of Analysis | 13/08/2024 | A15804.22,A15804.14,A15804.36 |
P05K003 | Certificate of Analysis | 25/03/2024 | A15804.22,A15804.14,A15804.36 |
Q12F017 | Certificate of Analysis | 05/03/2024 | A15804.22,A15804.14,A15804.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.