missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
£49.70 - £133.00
Chemical Identifiers
CAS | 137-66-6 |
---|---|
Molecular Formula | C22H38O7 |
Molecular Weight (g/mol) | 414.54 |
MDL Number | MFCD00005377 |
InChI Key | NULLRXHRYOXSEW-UHFFFAOYNA-N |
Synonym | ascorbyl palmitate, l-ascorbyl 6-palmitate, 6-o-palmitoyl-l-ascorbic acid, ascorbic palmitate, l-ascorbyl palmitate, unii-qn83us2b0n, ascorbyl monopalmitate, l-ascorbic acid 6-palmitate, s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate, ascorbyl palmitate nf |
PubChem CID | 54680660 |
IUPAC Name | [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate |
SMILES | CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15100337
|
Thermo Scientific Acros
449480250 |
25 g |
£49.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15214774
|
Thermo Scientific Acros
449481000 |
100 g |
£133.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
137-66-6 | |
414.54 | |
NULLRXHRYOXSEW-UHFFFAOYNA-N | |
54680660 | |
CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O |
C22H38O7 | |
MFCD00005377 | |
ascorbyl palmitate, l-ascorbyl 6-palmitate, 6-o-palmitoyl-l-ascorbic acid, ascorbic palmitate, l-ascorbyl palmitate, unii-qn83us2b0n, ascorbyl monopalmitate, l-ascorbic acid 6-palmitate, s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate, ascorbyl palmitate nf | |
[(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate |
Specifications
137-66-6 | |
250°C | |
Glass bottle | |
MFCD00005377 | |
Solubility in water: insoluble | |
CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O | |
414.54 | |
414.53 |
111°C to 115°C | |
0.1% max | |
C22H38O7 | |
ascorbyl palmitate, l-ascorbyl 6-palmitate, 6-o-palmitoyl-l-ascorbic acid, ascorbic palmitate, l-ascorbyl palmitate, unii-qn83us2b0n, ascorbyl monopalmitate, l-ascorbic acid 6-palmitate, s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate, ascorbyl palmitate nf | |
NULLRXHRYOXSEW-UHFFFAOYNA-N | |
[(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate | |
54680660 | |
L(+)-Ascorbyl palmitate |
Safety and Handling
missing translation for 'einecsNumber' : 205-305-4
RUO – Research Use Only