missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(-)-Carvone, 99%, Thermo Scientific Chemicals
£16.50 - £68.70
Chemical Identifiers
CAS | 6485-40-1 |
---|---|
Molecular Formula | C10H14O |
Molecular Weight (g/mol) | 150.22 |
MDL Number | MFCD00001578 |
InChI Key | ULDHMXUKGWMISQ-SECBINFHSA-N |
Synonym | --carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone |
PubChem CID | 439570 |
ChEBI | CHEBI:15400 |
IUPAC Name | (5R)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one |
SMILES | CC(=C)[C@@H]1CC=C(C)C(=O)C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10307620
|
Thermo Scientific Acros
154590050 |
5 g | Glass bottle |
£16.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10498120
|
Thermo Scientific Acros
154591000 |
100 g | Glass bottle |
£22.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041810
|
Thermo Scientific Acros
154595000 |
500 g | Glass bottle |
£68.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6485-40-1 | |
150.22 | |
ULDHMXUKGWMISQ-SECBINFHSA-N | |
439570 | |
(5R)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one |
C10H14O | |
MFCD00001578 | |
--carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone | |
CHEBI:15400 | |
CC(=C)[C@@H]1CC=C(C)C(=O)C1 |
Specifications
6485-40-1 | |
0.9580g/mL | |
88°C | |
98.5% min. (GC) | |
1.4950 to 1.5020 | |
07, 157 | |
0.958 | |
--carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone | |
ULDHMXUKGWMISQ-SECBINFHSA-N | |
(5R)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one | |
439570 | |
150.22 | |
Liquid |
Colorless to Yellow | |
227.0°C to 230.0°C | |
Authentic | |
C10H14O | |
MFCD00001578 | |
14, 1874 | |
− 57.00 | |
Solubility in water: practically insoluble in water. Other solubilities: soluble in alcohol, ether, chloroform,, propylene glycol and mineral oils, insoluble in glycerol | |
CC(=C)[C@@H]1CC=C(C)C(=O)C1 | |
150.22 | |
CHEBI:15400 | |
99% | |
L(-)-Carvone, 99+% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 229-352-5
missing translation for 'rtecsNumber' : OS8650000
missing translation for 'tsca' : TSCA
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- L(-)-Carvone
Signal Word
- Warning
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only