missing translation for 'onlineSavingsMsg'
Learn More
L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis), Thermo Scientific Chemicals Product Code.: 15895448

L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis), Thermo Scientific Chemicals

Product Code. 15895448 Shop All Thermo Scientific Products
100 g, Plastic Bottle
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 15895448

Brand: Thermo Scientific J14035.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7048-04-6
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hclShow MoreShow Less
PubChem CID 23462
IUPAC Name (2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Melting Point ∼175°C (decomposition)
Packaging Plastic Bottle
Linear Formula HSCH2CH(NH2)COOH.HCl.H2O
Beilstein 5158059
Quantity 100 g
Formula Weight 175.63
Percent Purity 98.5 to 101.5%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Cysteine Hydrochloride Monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number240346 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ14035.14,J14035.A7,J14035.22,J14035.A1
Lot Number200155 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ14035.14,J14035.A7,J14035.22,J14035.A1
Lot Number203590 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ14035.14,J14035.A7,J14035.22,J14035.A1
Lot Number233969 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberJ14035.14,J14035.A7,J14035.22,J14035.A1
Lot Number182777 Certificate TypeCertificate of Analysis Date27/11/2022 Catalog NumberJ14035.14,J14035.A7,J14035.22,J14035.A1
compliance-icons
Product Identifier
  • L-Cysteine hydrochloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : L-Cysteine hydrochloride monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.