Learn More
Thermo Scientific Chemicals L-Glutamic acid, 99%, Thermo Scientific Chemicals
Description
It is used as a chiral building-block in the synthesis of the (S)-isomer of -butyrolactone-4-carboxylic acid. L-Glutamic acid is widely used in fields of medicine, food processing, industry, etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a chiral building-block in the synthesis of the (S)-isomer of -butyrolactone-4-carboxylic acid. L-Glutamic acid is widely used in fields of medicine, food processing, industry, etc.
Solubility
It is very slightly soluble in cold water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.

Specifications
Specifications
Melting Point | 189°C to 192°C (decomposition) |
Assay Percent Range | ≥99% |
Beilstein | 1723801 |
Merck Index | 14,4469 |
Solubility Information | It is very slightly soluble in cold water. |
Density | 1.525 |
Quantity | 1000 g |
Formula Weight | 147.13 |
Optical Rotation | +31° (c=5 in 5N HCl) |
Percent Purity | ≥99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10254281 | Certificate of Analysis | 10/04/2025 | A15031.30,A15031.0B,A15031.36,A15031.0I |
10252642 | Certificate of Analysis | 11/10/2024 | A15031.30,A15031.0B,A15031.36,A15031.0I |
10247194 | Certificate of Analysis | 07/07/2023 | A15031.30,A15031.0B,A15031.36,A15031.0I,A15031 |
10149849 | Certificate of Analysis | 10/06/2022 | A15031.30,A15031.0B,A15031.36,A15031.0I |
10214043 | Certificate of Analysis | 10/06/2022 | A15031.30,A15031.0B,A15031.36,A15031.0I |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.