Learn More
Thermo Scientific Chemicals L-Histidine, 98+%
£18.80 - £226.00
Chemical Identifiers
CAS | 71-00-1 |
---|---|
Molecular Formula | C6H9N3O2 |
Molecular Weight (g/mol) | 155.16 |
MDL Number | MFCD00064315 |
InChI Key | HNDVDQJCIGZPNO-UHFFFAOYNA-N |
Synonym | l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin |
PubChem CID | 6274 |
ChEBI | CHEBI:15971 |
SMILES | NC(CC1=CN=CN1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418613
|
Thermo Scientific Chemicals
A10413.14 |
25 g |
£18.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428613
|
Thermo Scientific Chemicals
A10413.22 |
100 g |
£55.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438613
|
Thermo Scientific Chemicals
A10413.36 |
500 g |
£226.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.
Solubility
Soluble in water (15 mg/ml at 20°C).
Notes
Store at 4°C.Incompatible with oxidizing agents.
Chemical Identifiers
71-00-1 | |
155.16 | |
HNDVDQJCIGZPNO-UHFFFAOYNA-N | |
6274 | |
NC(CC1=CN=CN1)C(O)=O |
C6H9N3O2 | |
MFCD00064315 | |
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin | |
CHEBI:15971 |
Specifications
∽281°C (decomposition) | |
>98% | |
MFCD00064315 | |
14,4720 | |
Soluble in water (15mg/ml at 20°C). | |
NC(CC1=CN=CN1)C(O)=O | |
155.16 | |
CHEBI:15971 | |
≥98% |
71-00-1 | |
C6H9N3O2 | |
4673585 | |
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin | |
HNDVDQJCIGZPNO-UHFFFAOYNA-N | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid | |
6274 | |
155.16 | |
L-Histidine |
Safety and Handling
missing translation for 'einecsNumber' : 200-745-3
missing translation for 'rtecsNumber' : MS3070000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only