missing translation for 'onlineSavingsMsg'
Learn More
L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals Product Code.: 10444045

L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 10444045
500 g, Plastic Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10444045

Brand: Thermo Scientific Acros 411735000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrateShow MoreShow Less
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Ammonium 200ppm max.
pH 3 to 5 (2.5% soln. at 25°C)
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98%
Loss on Drying 7% to 10% (3 hrs) (145 to 150°C)
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Beilstein 25, 513
Specific Rotation + 9.90
Sulfate 300ppm max.
Merck Index 10, 4617
Sulfated Ash 0.1% max.
Specific Rotation Condition + 9.90 (20.00°C c=11,6 N HCl)
Quantity 500 g
Formula Weight 209.64
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Histidine hydrochloride monohydrate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465440 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number411730025,411735000,411730250,411731000
Lot NumberA0465176 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number411730025,411735000,411730250,411731000
Lot NumberA0463133 Certificate TypeCertificate of Analysis Date03/06/2024 Catalog Number411730025,411735000,411730250,411731000
Lot NumberA0462254 Certificate TypeCertificate of Analysis Date24/04/2024 Catalog Number411730025,411735000,411730250,411731000
Lot NumberA0459206 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number411730025,411735000,411730250,411731000
Product Identifier
  • L-Histidine hydrochloride monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.