missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L(+)-Lysine Monohydrochloride, ≥99% Product Code.: 10274930

Thermo Scientific Chemicals L(+)-Lysine Monohydrochloride, ≥99%

Product Code. 10274930
1 kg, Plastic Bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10274930

Brand: Thermo Scientific Chemicals 125220010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisylShow MoreShow Less
PubChem CID 69568
ChEBI CHEBI:53633
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 263.0°C to 264.0°C
Color White
Foreign Amino Acids 0.5% max. (TLC, 50μgspotting)
Heavy Metals (as Pb) 15ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99+%
Loss on Drying 0.4% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula H2N(CH2)4CH(NH2)CO2H·HCl
Specific Rotation + 21.00
Sulfate 300ppm max.
Merck Index 15, 5697
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 65 g/100 mL (20°C).
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrochloride
Specific Rotation Condition +21° (20°C c=8,6N HCl)
Quantity 1 kg
Formula Weight 182.65
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Lysine monohydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461045 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number125220010,125220050,125221000,125222500
Lot NumberA0450662 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number125220010,125220050,125221000,125222500
Lot NumberA0398271 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number125220010,125220050,125221000,125222500
Lot NumberA0417613 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number125220010,125220050,125221000,125222500
Lot NumberA0446006 Certificate TypeCertificate of Analysis Date29/09/2022 Catalog Number125220010,125220050,125221000,125222500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L(+)-Lysine Monohydrochloride, ≥99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.