missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Lysine monohydrochloride, 99+% Product Code.: 11497707

Thermo Scientific Chemicals L-Lysine monohydrochloride, 99+%

Product Code. 11497707
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11497707

Brand: Thermo Scientific Chemicals A16249.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine.  L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine.  L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point ∼260°C (decomposition)
Beilstein 3563889
Merck Index 14,5636
Solubility Information It is soluble in water.
Quantity 250 g
Formula Weight 182.65
Optical Rotation +20° (c=5 in 5N HCl)
Percent Purity ≥99%
Chemical Name or Material L-Lysine monohydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY15K062 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberA16249.18,A16249.30,A16249.0B
Lot Number10251955 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberA16249.18,A16249.30,A16249.0B
Lot NumberQ27K003 Certificate TypeCertificate of Analysis Date10/05/2024 Catalog NumberA16249.18,A16249.30,A16249.0B
Lot Number10249300 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog NumberA16249.18,A16249.30,A16249.0B
Lot NumberY17J006 Certificate TypeCertificate of Analysis Date03/01/2024 Catalog NumberA16249.18,A16249.30,A16249.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Lysine monohydrochloride, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.