missing translation for 'onlineSavingsMsg'
Learn More
L-(-)-Malic acid, 99%, Thermo Scientific Chemicals Product Code.: 15455479

L-(-)-Malic acid, 99%, Thermo Scientific Chemicals

Product Code. 15455479
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 15455479

Brand: Thermo Scientific Alfa Aesar J63221.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.

Solubility
Soluble in water(363g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-67-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064213
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
Synonym l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acidShow MoreShow Less
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2S)-2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Melting Point 100°C to 106°C
Color White
Density 1.6
Flash Point 220°C (428°F)
Quantity 100 g
Beilstein 1723541
Merck Index 14,5707
Solubility Information Soluble in water(363g/L).
Optical Rotation −26° (c=5.5 in Pyridine)
Formula Weight 134.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Malic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP24J028 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog NumberJ63221.36,J63221.A4,J63221.22
Lot NumberW27J005 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog NumberJ63221.36,J63221.A4,J63221.22
Lot NumberS24I030 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog NumberJ63221.36,J63221.A4,J63221.22
Lot NumberY27D005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63221.36,J63221.A4,J63221.22
Lot NumberP26B018 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63221.36,J63221.A4,J63221.22
compliance-icons
Product Identifier
  • L-(-)-Malic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : L-(-)-Malic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-(-)-Malic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.