missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Phenylalanine, 98.5-101.0%, Thermo Scientific Chemicals
£34.57 - £488.05
Chemical Identifiers
CAS | 63-91-2 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.19 |
InChI Key | COLNVLDHVKWLRT-UHFFFAOYNA-N |
Synonym | l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh |
PubChem CID | 6140 |
ChEBI | CHEBI:17295 |
SMILES | NC(CC1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15229886
|
Thermo Scientific Acros
453930250 |
25 g |
£34.57
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15239886
|
Thermo Scientific Acros
453931000 |
100 g |
£96.08
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15279846
|
Thermo Scientific Acros
453930010 |
1 kg | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63-91-2 | |
165.19 | |
l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
CHEBI:17295 |
C9H11NO2 | |
COLNVLDHVKWLRT-UHFFFAOYNA-N | |
6140 | |
NC(CC1=CC=CC=C1)C(O)=O |
Specifications
270.0°C to 275.0°C | |
5.4 to 6.0 (1% solution) | |
15ppm max. | |
Authentic | |
0.003% max. | |
l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
COLNVLDHVKWLRT-UHFFFAOYNA-N | |
2-amino-3-phenylpropanoic acid | |
165.19 | |
CHEBI:17295 | |
165.19 | |
−34.10 (20.00°C c=2,water,dry basis) |
63-91-2 | |
0.05% max. | |
C9H11NO2 | |
C6H5CH2CH(NH2)CO2H | |
0.3% max. (105°C, 3 hrs) | |
Solubility in water: 27g/L (20°C). Other solubilities: very slightly soluble in methanol, ethanol | |
NC(CC1=CC=CC=C1)C(O)=O | |
−34.10 | |
6140 | |
0.03% max. | |
0.4% max. | |
L-Phenylalanine, 98.5-101.0% |
RUO – Research Use Only