missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Phenylalanine, 98.5+%
£23.92 - £1058.27
Chemical Identifiers
CAS | 63-91-2 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.19 |
MDL Number | MFCD00064227 |
InChI Key | COLNVLDHVKWLRT-UHFFFAOYNA-N |
Synonym | l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh |
PubChem CID | 6140 |
ChEBI | CHEBI:17295 |
SMILES | NC(CC1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10574061
|
Thermo Scientific Chemicals
130310250 |
25 g | Plastic Bottle |
£23.92
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10584061
|
Thermo Scientific Chemicals
130311000 |
100 g | Plastic Bottle |
£62.51
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10445175
|
Thermo Scientific Chemicals
130315000 |
500 g | Plastic Bottle |
£226.13
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13438348
|
Thermo Scientific Chemicals
130310025 |
2.5 kg | Plastic Bottle |
£1058.27
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63-91-2 | |
165.19 | |
COLNVLDHVKWLRT-UHFFFAOYNA-N | |
6140 | |
NC(CC1=CC=CC=C1)C(O)=O |
C9H11NO2 | |
MFCD00064227 | |
l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
CHEBI:17295 |
Specifications
270.0°C to 275.0°C | |
5.4 to 6.0 (1% aq. soln.) | |
20ppm max. | |
C9H11NO2 | |
C6H5CH2CH(NH2)CO2H | |
10ppm max. | |
l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
COLNVLDHVKWLRT-UHFFFAOYNA-N | |
2-amino-3-phenylpropanoic acid | |
165.19 | |
CHEBI:17295 | |
≥98.5% | |
− 34.10 (20.00°C c=2,water,dry basis) | |
L-Phenylalanine |
63-91-2 | |
White | |
98.5+% | |
Authentic | |
MFCD00064227 | |
0.3% max. (105°C, 3 hrs) | |
Solubility in water: 27g/L (20°C). Other solubilities: very slightly soluble in methanol, ethanol | |
NC(CC1=CC=CC=C1)C(O)=O | |
− 34.10 | |
6140 | |
165.19 | |
0.1% max. | |
Crystalline Powder or Crystals |
RUO – Research Use Only