Learn More
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assembly
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assemblies extend the operating life of standard pump heads
Brand:  Masterflex™ 07014-81
 View more versions of this product
Product Code. 11765168
  • £116.00 / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement parts to keep pump head operating for an extended period of time
  • Two end bell assemblies are required for a complete head assembly
  • Choose replacement parts suitable for your I/P™ Standard pump head size
  • Suitable for use with L/S™ Standard Pump Heads
Specifications
Specifications
CRS Rotor, L/S 14 Tubing
L/S™ Standard Pump Heads 07014-00, 07014-20, 07014-70, and 07014-72
Replacement End Bell Assembly
Polycarbonate
L/S 14