Learn More
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assembly
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assemblies extend the operating life of standard pump heads
£106.00 - £142.00
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
14821302
|
Masterflex™
07024-81 |
CRS Rotor, L/S 24 Tubing |
£106.00
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
11875202
|
Masterflex™
07014-91 |
SS Rotor, L/S 14 Tubing |
£142.00
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
11765168
|
Masterflex™
07014-81 |
CRS Rotor, L/S 14 Tubing |
£106.00
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
12182120
|
Masterflex™
07015-81 |
CRS Rotor, L/S 15 Tubing |
£107.00
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
11854252
|
Masterflex™
07016-81 |
CRS Rotor, L/S 16 Tubing |
£106.00
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
15217911
|
Masterflex™
07018-81 |
CRS Rotor, L/S 18 Tubing |
£106.00
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
|||||
Description
- Replacement parts to keep pump head operating for an extended period of time
- Two end bell assemblies are required for a complete head assembly
- Choose replacement parts suitable for your I/P™ Standard pump head size
- Suitable for use with L/S™ Standard Pump Heads
Specifications
Polycarbonate | |
Replacement End Bell Assembly |