missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Serinamide hydrochloride, 98%, Thermo Scientific™
£17.70 - £35.50
Chemical Identifiers
CAS | 65414-74-6 |
---|---|
Molecular Formula | C3H9ClN2O2 |
Molecular Weight (g/mol) | 140.567 |
MDL Number | MFCD00067560 |
InChI Key | VURWDDZIWBGXCK-DKWTVANSSA-N |
Synonym | l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl |
PubChem CID | 16212403 |
IUPAC Name | (2S)-2-amino-3-hydroxypropanamide;hydrochloride |
SMILES | C(C(C(=O)N)N)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10473035
|
Thermo Scientific Acros
303162500 |
250 mg | Glass Bottle |
£17.70
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073550
|
Thermo Scientific Acros
303160010 |
1 g | Glass Bottle | ||||||
Description
Chemical Identifiers
65414-74-6 | |
140.567 | |
VURWDDZIWBGXCK-DKWTVANSSA-N | |
16212403 | |
C(C(C(=O)N)N)O.Cl |
C3H9ClN2O2 | |
MFCD00067560 | |
l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl | |
(2S)-2-amino-3-hydroxypropanamide;hydrochloride |
Specifications
184°C to 190°C | |
Purple to Red | |
C3H9ClN2O2 | |
HOCHCH(NH)CONH·HCl | |
l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl | |
C(C(C(=O)N)N)O.Cl | |
+13.5° to +15.5° (20°C, 589nm) (c=1, H2O) | |
16212403 | |
≥97.5% (Argentometry) | |
Crystalline Powder or Crystals |
65414-74-6 | |
98% | |
Authentic | |
MFCD00067560 | |
VURWDDZIWBGXCK-DKWTVANSSA-N | |
(2S)-2-amino-3-hydroxypropanamide;hydrochloride | |
140.567 | |
140.57 | |
+14.5° (20°C c=1,H2O) | |
L-Serinamide hydrochloride |
RUO – Research Use Only