missing translation for 'onlineSavingsMsg'
Learn More

Techsit™ L-Tech Laboratory High Stool

Antimicrobial vinyl laboratory stool is fully adjustable with an adjustable foot ring.

Brand:  Techsit™ TS8341B

Encompass_Preferred

 View more versions of this product

Product Code. 16553934

  • £325.42 / Each
Estimated Shipment: 21-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Vinyl high stool with adjustable foot ring
  • Black Antimicrobial Vinyl that is easy to clean
  • Fully adjustable seat height
  • Cleanroom Class 7 suitable
Specifications

Specifications

Laboratory Stool
Yes
Vinyl
Gas Lift
High Bench Height
5 Year
Glide
No
No
No
Vinyl
Black
Not Autoclavable
Steel
150 kg
Vinyl
16 kg
400 mm diameter
No
Yes
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers