Learn More
Eagle Lab Packs with Screw-On Lid
Eagle Lab Packs are ideal for safe storage, handling and transportation of hazardous materials, spill containment and clean-up of hazardous materials, original shipping container, bio waste disposal and incineration.
Brand: Eagle EDR1600SYL
66.06 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Eagle Lab Pack Drums are constructed of Yellow blow-molded, high-density polyethylene (HDPE) with UV inhibitors, are durable and lightweight, weather and chemical resistant, and screw-on lid
- Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
- Lab Packs/Open Head Drums- for use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials - if leaking, must pack with sufficient sorbents for - NO FREE LIQUIDS
Specifications
113 L | |
Yellow | |
Safe Storage, Handling and Transportation of Hazardous Materials | |
Polyethylene |