Learn More
Epredia™ Lab Vision™ biotinylated Goat anti-Polyvalent (Ready-To-Use)
Employ a labeled streptavidin-biotin immunoenzymatic antigen detection system with the Epredia™ Lab Vision™ Biotinylated Goat anti-Polyvalent (Ready-To-Use).
Brand: Epredia™ TP-125-BN
Code : LD
Additional Details : Weight : 0.18144kg
Description
This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate-chromogen.Species: Goat
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Conjugate: Biotin
Specifications
Biotin | |
Anti-Polyvalent | |
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
2° to 8°C | |
125mL |