missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ biotinylated Goat anti-Polyvalent (Ready-To-Use)

Employ a labeled streptavidin-biotin immunoenzymatic antigen detection system with the Epredia™ Lab Vision™ Biotinylated Goat anti-Polyvalent (Ready-To-Use).

£373.00 - £575.00

Specifications

Conjugate Biotin
Content And Storage 2° to 8°C
Product Type Anti-Polyvalent
Species anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12674047
View Documents
Epredia™
TP-060-BN
60 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12624057
View Documents
Epredia™
TP-125-BN
125mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate-chromogen.
  • Species: Goat

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Conjugate: Biotin

Specifications

Specifications

Biotin
2° to 8°C
Anti-Polyvalent
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 18 Months

SDS
Documents

Documents

Product Certifications