missing translation for 'onlineSavingsMsg'
Learn More

Calprtectin-Monocyte/Macrophage/L1 protein Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ Calprtectin-Monocyte/Macrophage/L1 protein Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-148-P

286.00 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.


 View more versions of this product

Product Code. 12639916

  • £329.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Mouse

Clone: MAC 387

Isotype: IgG1

Species Reactivity: Human, Baboon, Monkey, Pig, Cat, Dog, Guinea pig, Rat, and Mouse. Others not known.

Epitope: Not determined

Immunogen: Affinity purified monocyte membrane preparation

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Macrophage/L1 Protein/Calprotectin Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
Hematopoietic
Canine, Feline, Guinea Pig, Human, Mouse, Non-human Primate, Porcine, Rat
IgG1
Immunohistochemistry (Paraffin)
MAC 387
Unconjugated
Affinity purified monocyte membrane preparation
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.