Learn More
Calprtectin-Monocyte/Macrophage/L1 protein Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Calprtectin-Monocyte/Macrophage/L1 protein Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
£98.43 - £364.00
Specifications
Antigen | Macrophage/L1 Protein/Calprotectin Ab-1 |
---|---|
Clone | MAC 387 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12649916
|
Epredia
MS-148-P0 |
Concentrated | 100 μL | N/A | |||||
12669916
|
Epredia
MS-148-P1ABX |
Concentrated | 100 μL | N/A | |||||
12679916
|
Epredia
MS-148-PABX |
Concentrated | 200 μL | N/A | |||||
12659916
|
Epredia
MS-148-P1 |
Concentrated | 500 μL | N/A | |||||
12639916
|
Epredia
MS-148-P |
Concentrated | 1 mL | N/A | |||||
12689916
|
Epredia
MS-148-R7 |
Prediluted | 7 mL | N/A | |||||
Description
Host Species: Mouse
Clone: MAC 387
Isotype: IgG1
Species Reactivity: Human, Baboon, Monkey, Pig, Cat, Dog, Guinea pig, Rat, and Mouse. Others not known.
Epitope: Not determined
Immunogen: Affinity purified monocyte membrane preparation
Positive Control: Tonsil, lymph node, or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Macrophage/L1 Protein/Calprotectin Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Canine, Feline, Guinea Pig, Human, Mouse, Non-human Primate, Porcine, Rat | |
IgG1 |
MAC 387 | |
Monoclonal | |
Mouse | |
Hematopoietic | |
Affinity purified monocyte membrane preparation | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.