missing translation for 'onlineSavingsMsg'
Learn More

Estrogen Receptor (SP1) Rabbit Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Estrogen Receptor (SP1), Rabbit Monoclonal Antibody.

Brand:  Epredia RM-9101-S0

 View more versions of this product

Product Code. 12623667

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Recognizes a protein of 67 kDa, which is identified as estrogen receptor (ER) alpha. The ER gene consists of more than 140kb of genomic DNA divided into 8 exons, and is translated into a protein with six functionally discrete domains, labeled A through F. This antibody strongly stains the nucleus of epithelial cells in breast carcinomas. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Clone: SP1

Isotype: IgG

Species Reactivity: Human, Mouse and Hamster. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide derived from C-terminal of human estrogen receptor

Molecular Weight: 67 kDa

Positive Control: Breast CA, MCF-7 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence (Not verified)
  • Flow Cytometery (Not verified)
Specifications

Specifications

Estrogen Receptor (SP1)
Monoclonal
Unconjugated
Synthetic peptide derived from C-terminal of human estrogen receptor
IVD
Primary
Concentrated
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
SP1
Rabbit
100 μL
Steroids Hormones and Growth Factors
Hamster, Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Estrogen Receptor (SP1) Rabbit Monoclonal Antibody, Epredia™ > 100μL; Unlabeled; Supernatant

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.