missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ PermaFluor™ Aqueous Mounting Medium
Enhance FITC intensity in fluorescence microscopy with Epredia™ Lab Vision™ PermaFluor™ Aqueous Mounting Medium.
Brand: Epredia™ TA-030-FM
180.96 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reduces fading
- Immobilizes cover glass over tissue section
Storage Conditions: Store at 2°-8°C
Specifications
Store at 2° to 8°C | |
30 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction