missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X)

Improve the reactivity of certain antibodies in formalin-fixed tissues with Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X).

Brand:  Epredia™ AP-9005-125

 View more versions of this product

Product Code. 12688286

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 100X stock solution
  • 100mM Tris-HCl buffer, pH 10.0
  • Dilute 10-fold with distilled water before use

Storage Conditions: Store at 2-8°C. Stable for up to 18 months. Contains no preservatives.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C.
HIER
Immunohistochemistry
10X
125mL
10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X) > 125mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.