missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Microcentrifuge Tube Racks

Hold tubes upright and promote conduction of heat or cold from Temp-Zone area on selected models of Purifier Cell Logic Biosafety Cabinets to help maintain sample temperature

£134.00 - £321.00

Specifications

Product Type Tube Rack
Autoclavable Not Autoclavable
Color Blue
Material Aluminum
View More Specs

Products 4
Product Code Brand Dimensions (L x W x H) Positions Holds Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Positions Holds Price Quantity & Availability  
15989501
View Documents
Labconco™
4026403
8 x 8 x 1.5 in. (20.3 x 20.3 x 3.8cm) 96 Microtubes (1.5 mL)
£321.00
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
16358887
View Documents
Labconco™
4026401
8 x 6 x 1.5 in. (20.3 x 15.2 x 3.8cm) 96 Microtubes (0.5 mL)
£296.00
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
16802566
View Documents
Labconco™
4026402
6 x 2 x 1.5 in. (15.2 x 5.1 x 3.8cm) 18 Microtubes (1.5 mL)
£191.00
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
15546029
View Documents
Labconco™
4026405
5 x 2 x 2.5 in. (12.7 x 5.1 x 6.4cm) 12 Tubes (5 mL)
£134.00
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
Description

Description

Hold tubes upright and promote conduction of heat or cold from Temp-Zone area on selected models of Purifier Cell Logic Biosafety Cabinets to help maintain sample temperature

  • Material: Anodized aluminum
  • Color: Blue

Specifications

Specifications

Tube Rack
Not Autoclavable
Blue
Aluminum
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.