Learn More
Labconco™ Protector XVS™ Flexible Duct Connection for Ventilation Stations
Flexible Duct Connection for Ventilation Stations
Brand: Labconco™ 4861800
This item is not returnable.
View return policy
Description
- For use with Labconco Protector XVS Ventilation Stations
- Flexible duct connection reduces vibration between blower and PVC ductwork; supplied with two installation clamps
Specifications
Protector VS Ventilation Station | |
8 lb. | |
30.48 cm | |
6 in. dia. round PVC duct | |
Ducting |
12 in. | |
3.6 Kg | |
Flexible Duct Connection | |
PVC |
Product Content Correction
Product Title
Labconco™ Protector XVS™ Flexible Duct Connection for Ventilation Stations > Flexible PVC Duct Connection
Spot an opportunity for improvement?Share a Content Correction